Skip to Content
Merck
All Photos(1)

Key Documents

Z678813

2mag bioMIXdrive 4 magnetic stirrer

for use with bioMIXcontrol S control unit

Synonym(s):

induction multi-position stirrer, magnetic induction stir plate, magnetic wear-free stirrer, multi-position stirrer

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€2,300.00

€2,300.00


Check Cart for Availability


Select a Size

Change View
1 EA
€2,300.00

About This Item

UNSPSC Code:
41103806
NACRES:
NB.72

€2,300.00


Check Cart for Availability

material

stainless steel plate

feature

CE compliant
positions 4

packaging

pack of 1 ea

manufacturer/tradename

2mag 80004

parameter

5-250 rpm speed

plate size × W × L

× 270 mm × 270 mm

stirrer volume

5-5000 mL , per stirring position

Looking for similar products? Visit Product Comparison Guide

General description

2mag bioMIXcontrol S unit is needed, sold separately.
Warming and maintenance-free magnetic stirrer with 4 stirring points with large surface, up to 5,000 mL stirring volume per point, also suitable for viscous culture media.

Maintenance-free 2mag-stepper-motor-drive for extremely slow and 100% jerk-free and gentle mixing of cell cultures, extended speed range from 5 up to 250 rpm, jerk-free, even and smooth stirring also at slow speeds, suitable for culture flasks with spinning propeller- and spinning ball systems.

Fully encapsulated and hermetically closed stainless steel housing, IP68, water-, dust- and germ-proof, extremely robust construction and performance, easy to clean and maximum firm, submersible in water baths, designed for the warming-free long term operation in CO2-incubators, individual measurements on request.

2mag bioMIXcontrol S unit is needed, sold separately.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service