Skip to Content
Merck
All Photos(1)

Documents

Z676179

Plum neutralizing and saline eye wash station

contains 200 mL sterile pH neutralizing solution and 500 mL sterile saline eyewash

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

manufacturer/tradename

Plum

General description

Effective rinsing solution for removing foreign bodies & neutralisation of acids / alkalis. This open eye wash station contains 200 mL sterile pH Neutralizing solution and 500 mL sterile saline eyewash.

Especially suitable for workplaces where there is a risk of injuries from acids or alkalis. The station is equipped with a mirror and clear instructions for use. Each bottle has an ergonomic eyecup that fits the eye and ensures a gentle, even rinsing.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service