Skip to Content
Merck
All Photos(1)

Key Documents

Z674680

Slide spinner for slide drying

AC/DC input 230 V AC, UK plug

Synonym(s):

slide dryer

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€607.00

€607.00


Check Cart for Availability


Select a Size

Change View
1 EA
€607.00

About This Item

UNSPSC Code:
41100000
NACRES:
MA.01

€607.00


Check Cart for Availability

manufacturer/tradename

Labnet C1303-T-230V-UK

parameter

~4800 rpm speed

W × D × H

5 in. (12.7 cm) × 5 in. (12.7 cm) × 4.4 in. (11.2 cm)

Looking for similar products? Visit Product Comparison Guide

General description

Simply insert your slides into the cassettes and close the lid. This Slide Spinner quickly accelerated to 4800 RPM, the optimal speed for quick drying without any splashing. Open the lid and the rotor comes to a stop and the dry slides can be removed. The cassettes catch all of the liquid, and can easily be removed, and the liquid can be poured out.
  • Quickly dries 2 slides
  • Ideal for removing wash buffer from microarrays prior to scanning
Removeable cassettes conveniently catch all liquid

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service