Z674141
Duran® GLS 80 wide mouth graduated laboratory bottles with cap and pouring ring
capacity 20,000 mL
Synonym(s):
lab bottle, lab bottles, wide mouth glass
Select a Size
€770.00
Select a Size
About This Item
€770.00
Recommended Products
material
glass bottle
polypropylene cap
round bottle
Joint
Joints threaded neck (GLS80)
sterility
non-sterile
feature
graduations
with cap
packaging
pack of 1 ea
manufacturer/tradename
DURAN® 1113951
capacity
20,000 mL
neck I.D.
80 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Duran glass elliminates ion leaching problems
- Sturdy design enhances safety and service life
- Thermal properties allow sterilisation by autoclave
- Convenient and easy to read graduations
- Supplied with blue quick-release cap and pouring ring that is autoclavable up to 140°C
Features and Benefits
- Manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Special thread design for rapid opening and closing in less than a turn
- Optimum filling and pouring and particularly easy cleaning
- Spoons, spatulas and tweezers can be manipulated with ease
- Even large-volume funnels can be inserted into the bottles without any difficulty
- Stability due to the large base area
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service