Skip to Content
Merck
All Photos(1)

Documents

Z561754

Coors high alumina combustion boat

capacity 20 mL

Synonym(s):

Combustion Boat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.43

packaging

pack of 1 ea

manufacturer/tradename

Coorstek 65566

L × W × H

105 mm × 22 mm × 14.5 mm

capacity

20 mL

Looking for similar products? Visit Product Comparison Guide

Application

Coors high alumina combustion boat has been used in the preparation of gold (Au) substrates for use in a complementary “conductance plateau” technique, the scanning tunneling microscopy (STM) current-distance (I(s)) experiment.

Legal Information

Coors is a trademark of Coorstek, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service