Key Documents
Z532088
Sigma-Aldrich
Solid glass plug
O.D. 1/2 in.
Select a Size
1 EA
€23.50
€23.50
Check Cart for Availability
Select a Size
Change View
1 EA
€23.50
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
€23.50
Check Cart for Availability
O.D.
1/2 in.
12.7 mm
General description
Replace transfer tubing with this plug to make a leakproof Torion seal.
Legal Information
Torion is a trademark of Bibby Sterlin Ltd.
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)