Key Documents
Z510831
Sigma-Aldrich
Connecting Adapter
female joint: ST/NS 24/40, thread size 20-400
Synonym(s):
747130-2024
Select a Size
1 EA
€59.20
€59.20
Check Cart for Availability
Select a Size
Change View
1 EA
€59.20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€59.20
Check Cart for Availability
thread size
20-400
Joint
female joint: ST/NS24/40
General description
Connects ground joint glassware to threaded microscale component or sample vials. Excellent for use with splash-guards to recover small quantities of material during rotary evaporation. Order threaded vial connectors separately.
Features and Benefits
  • Accessory for microscale MICROFLEX threaded kits
  • Connects Standard Taper outer ground joint to GPI thread of inner ground joint
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Choose from one of the most recent versions:
Certificates of Analysis (COA)