Key Documents
Z509337
Sigma-Aldrich
Weighing funnel
size S, anti-static PP
Synonym(s):
Weighing Apparatus, Weighing Device
Select a Size
50 EA
€85.50
€85.50
Check Cart for Availability
Select a Size
Change View
50 EA
€85.50
About This Item
UNSPSC Code:
41121809
NACRES:
NB.71
€85.50
Check Cart for Availability
material
anti-static PP
size
S
packaging
pkg of 50 ea
General description
  • Sit flat on balance - No rolling, no spilling!
  • Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer
  • Residual sample can be rinsed to guarantee 100% transfer
  • Suitable for safe handling of hazardous materials
  • Available in five different sizes: extra small, small, medium, large, and extra large
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Are these funnels autoclavable?
    1 answer
    1. While polypropylene is autoclavable, the anti-static agent applied to these funnels may contribute to degradation at high temperature and pressure.
      Autoclaving of this product has not been validated.
      Helpful?