Skip to Content
Merck
All Photos(1)

Key Documents

Z409472

Sigma-Aldrich

Weighing funnel

size S, polypropylene

Synonym(s):

Laboratory funnel, Weighing device

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
€61.40

€61.40


Check Cart for Availability


Select a Size

Change View
50 EA
€61.40

About This Item

UNSPSC Code:
41121809
NACRES:
NB.71

€61.40


Check Cart for Availability

material

polypropylene

size

S

packaging

pkg of 50 ea

Looking for similar products? Visit Product Comparison Guide

General description

  • Sit flat on balance - No rolling, no spilling!
  • Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer
  • Residual sample can be rinsed to guarantee 100% transfer
  • Suitable for safe handling of hazardous materials
  • Available in five different sizes: extra small, small, medium, large, and extra large

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. what is the weighing range of the size 's' weighing funnels?

    1 answer
    1. This product is a small-capacity weighing funnel. While there is no range stated, the reservoir maximum volume is 2 mL. Likewise, assuming a density of 1 g/mL, the maximum weight is 2 grams. Note that the maximum weight may not apply to very high or very low-density materials.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service