Z509027
AldraSORB™ water trapping packets
volume 1,000-4,000 mL, large
Synonym(s):
Humidity Control Packets
Select a Size
€122.00
Estimated to ship on28 May 2025
Select a Size
About This Item
€122.00
Estimated to ship on28 May 2025
Recommended Products
capacity
1000-4000 mL
volume
1,000-4,000 mL
weight
20 g
General description
- Maintains low levels of water impurities for up to 3 months
- Ideal for solid-phase and solution-phase peptide synthesis
- Effectively dries solvents to below 50 ppm
Note: These traps are not recommended for solvents having more than 0.05% or 500 ppm of water. They should not be used in acidic solvents.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)