Key Documents
Z511706
Sigma-Aldrich
AldraAmine trapping packets
volume 1,000-4,000 mL, large
Synonym(s):
AldraAmine Packets, AldraAmine Packs, Amine Trapping Packets
Select a Size
1 EA
€27.90
5 EA
€122.00
25 EA
€445.00
€27.90
Estimated to ship on25 April 2025
Select a Size
Change View
1 EA
€27.90
5 EA
€122.00
25 EA
€445.00
About This Item
UNSPSC Code:
24141516
NACRES:
NB.43
€27.90
Estimated to ship on25 April 2025
capacity
1000-4000 mL
volume
1,000-4,000 mL
General description
Two common solvents used in peptide synthesis are dimethylformamide (DMF) and 1-methyl-2-pyrrolidinone (NMP). Both solvents will degrade naturally over time producing unwanted amine impurities that can contaminate your synthesis. AldraAmine trapping packets offer a solution to this problem by effectively reducing amine and water contaminants to levels near those of freshly distilled solvent. Just insert a packet into the bottle of solvent and let it do the rest. Packets are made of an ultra-high-molecular-weight polyethylene membrane that has been tested for safe long-term exposure to DMF and NMP. Packets are vacuum sealed.
  • Effectively reduces amine and water impurities for up to 3 months
  • Each packet individually vacuum sealed and fully activated for immediate use
  • Safe for automated synthesizers
  • Ideal for solid-phase and solution-phase peptide synthesis
Note: These traps are not designed to be used in aqueous solutions (greater than 500 ppm of water). Avoid exposure to solvents with a very high water content as heat may be produced.
Choose from one of the most recent versions:
Certificates of Analysis (COA)