Skip to Content
Merck
All Photos(1)

Key Documents

Z420212

Sigma-Aldrich

Spinbar® disposable magnetic stirring bar

size 1/2 in. × 1/8 in.

Synonym(s):

Stir Bar, Stirbar, Stirring Bar

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€140.00

€140.00


Check Cart for Availability


Select a Size

Change View
1 PKG
€140.00

About This Item

UNSPSC Code:
41103806
NACRES:
NB.72

€140.00


Check Cart for Availability

material

PTFE-coated
polygon PTFE-coated

packaging

pkg of 100 ea

manufacturer/tradename

Bel-Art F37112-0012

size

1/2 in. × 1/8 in.

Looking for similar products? Visit Product Comparison Guide

General description

For single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.

Application

Spinbar® disposable magnetic stirring bar is suitable for single-use applications to avoid contamination or for radioactive reagents mixing.

Features and Benefits

Spinbar® disposable magnetic stirring bar provides the following benefits:
  • Autoclave-friendly
  • Multifaceted magnetic bar surface offers high turbulance.

Legal Information

Spinbar is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service