Key Documents
Z300896
Quickfit® Reduction Adapters
top female joint: ST/NS 14/23, bottom male joint: ST/NS 29/32
Synonym(s):
DA14
Select a Size
1 EA
€27.60
€27.60
Check Cart for Availability
Select a Size
Change View
1 EA
€27.60
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€27.60
Check Cart for Availability
Joint
female joint: ST/NS top14/23
male joint: ST/NS bottom29/32
manufacturer/tradename
Quickfit DA 14
availability
not available in USA and Canada
General description
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)