Skip to Content
Merck
All Photos(1)

Documents

Z203440

Sigma-Aldrich

Aldrich® antisplash splash-guard adapter without return hole

female joint: ST/NS 29/32, male joint: ST/NS 29/32, capacity 100 mL

Synonym(s):

Splash guard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.35

Joint

female joint: ST/NS29/32
male joint: ST/NS29/32

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

General description

Aldrich® antisplash splash-guard adapter without return hole are traps that greatly reduce carry-over from foaming or bumping. Whatever comes up into this trap will stay and not go back to the evaporator flask.

Application

Aldrich® antisplash splash-guard adapter without return hole is ideal for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping".

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service