Key Documents
Z274887
Maintenance kit for KNF Laboport® solid PTFE vacuum pump
Maintenance kit for 34 Lpm pump
Select a Size
1 KIT
€262.00
€262.00
Check Cart for Availability
Select a Size
Change View
1 KIT
€262.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
€262.00
Check Cart for Availability
manufacturer/tradename
KNF
General description
Includes replacement PTFE diaphragm, head seals and two Kalrez valves.
Kits include replacement PTFE diaphragm, head seals, and two Kalrez® valves. Note: Two kits are required when servicing both heads of vacuum pump.
Other Notes
Two kits are required when servicing both heads of vacuum pump.
Legal Information
Kalrez is a registered trademark of E. I. du Pont de Nemours and Company
Laboport is a registered trademark of KNF Neuberger GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)