Skip to Content
Merck
All Photos(1)

Documents

Z273600

Apiezon® grease

T

Synonym(s):

Apiezon Grease, High-Performance Grease

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
15121900
NACRES:
NB.35

vapor pressure

5 × 10-9 mmHg ( 20 °C)

description

usage: high temp

manufacturer/tradename

Apiezon

parameter

+10-+120 °C temperature

weight

25 g

Looking for similar products? Visit Product Comparison Guide

General description

For low vapor pressure sealing and lubrication in high vacuum applications.

Legal Information

Apiezon is a registered trademark of M & I Materials Ltd.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

419.0 °F - closed cup

Flash Point(C)

215 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service