Key Documents
Z247723
Coors high-alumina crucible cover
diam. 39.4 mm
Select a Size
1 EA
€48.30
€48.30
Estimated to ship on12 April 2025
Select a Size
Change View
1 EA
€48.30
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
€48.30
Estimated to ship on12 April 2025
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 65608
diam.
39.4 mm
General description
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
For Z247626
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)