Skip to Content
Merck
All Photos(1)

Key Documents

Z242721

Sigma-Aldrich

Aldrich® cuvette washer

Synonym(s):

cuvette cleaning

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€903.00

€903.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€903.00

About This Item

UNSPSC Code:
41121813
NACRES:
SB.32

€903.00


Check Cart for Availability

Request a Bulk Order

technique(s)

UV/Vis spectroscopy: suitable

General description

High-capacity, 100 mL solvent reservoir allows less frequent filling. Rubber cushion seals cuvette to washer and prevents chipping. Borosilicate glass with vacuum inlet and 24/40 bottom joint.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service