Key Documents
Z214639
Microscale vacuum adapter
with drip-tip
Select a Size
1 EA
€55.00
€55.00
Estimated to ship on28 May 2025
Select a Size
Change View
1 EA
€55.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
€55.00
Estimated to ship on28 May 2025
manufacturer/tradename
Ace Glass 526106
General description
#7 Ace-Thred at top with a nylon bushing and FETFE® O-ring that form a compression seal with microscale condenser tube, filter funnel, bleed tube, or thermometer (6.5 to 7.0 mm diam.). Has bottom 14/10 inner joint and side hose connection. (Shown with Hirsch funnel sold separately.)
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)