Key Documents
Z186740
Gas bag
capacity, 1 gal
Synonym(s):
gas collection bag, gas collection vessel, gas sampling equipment
Select a Size
1 EA
€92.60
€92.60
Check Cart for Availability
Select a Size
Change View
1 EA
€92.60
About This Item
UNSPSC Code:
24111500
NACRES:
NB.51
€92.60
Check Cart for Availability
feature
capacity 1 gal
packaging
pack of 1 ea
General description
Gas bag made of heavy natural rubber suitable for collecting and storing large samples of gases. Commonly used with PVC stopcocks with a 13 mm OD such as Z186767.
Choose from one of the most recent versions:
Certificates of Analysis (COA)