Key Documents
Z156531
Sigma-Aldrich
Pinch clamps for SJ joints
joint: SJ 65/40
Select a Size
1 EA
€147.00
€147.00
Check Cart for Availability
Select a Size
Change View
1 EA
€147.00
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
€147.00
Check Cart for Availability
material
stainless steel
Joint
joint: SJ65/40
General description
For use with Z13,730-8 and Z13,731-6
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.
Choose from one of the most recent versions:
Certificates of Analysis (COA)