Key Documents
Z152412
Stainless steel 304 syringe needle
gauge 18, L 36 in.
Select a Size
1 EA
€157.00
€157.00
Check Cart for Availability
Select a Size
Change View
1 EA
€157.00
About This Item
UNSPSC Code:
41122002
NACRES:
NB.03
€157.00
Check Cart for Availability
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science
L
36 in.
gauge
18
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)