Key Documents
Z102717
Stainless steel 304 syringe needle
gauge 18, L 6 in.
Synonym(s):
Stainless Needle, Steel Syringe
Select a Size
1 EA
€43.10
€43.10
Check Cart for Availability
Select a Size
Change View
1 EA
€43.10
About This Item
UNSPSC Code:
42142600
eCl@ss:
32070500
NACRES:
NB.03
€43.10
Check Cart for Availability
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science 4195
L
6 in.
gauge
18
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)