Skip to Content
Merck
All Photos(1)

Key Documents

Z126039

Sigma-Aldrich

ACE Stir-Lube® stirrer lubricant

1 oz or 4 oz sizes

Sign Into View Organizational & Contract Pricing

Select a Size

1 OZ
€30.50
4 OZ
€97.40

€30.50


Check Cart for Availability


Select a Size

Change View
1 OZ
€30.50
4 OZ
€97.40

About This Item

UNSPSC Code:
15121900
NACRES:
NB.72

€30.50


Check Cart for Availability

manufacturer/tradename

Ace Glass 811710

General description

Low-melting. Only very thin film needed. A non-cooled ACE bearing can be operated at 1500 r.p.m. and water-cooled bearings up to 2000 r.p.m. for many hours with negligible wear.

Legal Information

ACE Stir-Lube is a registered trademark of Ace Glass, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service