Skip to Content
Merck
All Photos(1)

Key Documents

Z113964

Sigma-Aldrich

Wheaton glass serum bottle

volume 5 mL, clear, O.D. 20 mm, diam. × H 23 mm × 47 mm

Sign Into View Organizational & Contract Pricing

Select a Size

288 EA
€275.00

€275.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
288 EA
€275.00

About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

€275.00


Check Cart for Availability

Request a Bulk Order

material

borosilicate glass bottle

description

USP Type I

feature

autoclavable
closure type crimp top vial

O.D.

20 mm

capacity

5 mL

diam. × H

23 mm × 47 mm

volume

5 mL

color

clear

fitting

opening for 20 mm

Looking for similar products? Visit Product Comparison Guide

General description

It is autoclavable, easy sterilization, is used to contain substances to prevent contamination.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service