Key Documents
Z113034
Stainless steel 304 syringe needle
gauge 20, L 2 in.
Select a Size
1 EA
€52.80
€52.80
Check Cart for Availability
Select a Size
Change View
1 EA
€52.80
About This Item
UNSPSC Code:
42142600
eCl@ss:
32070500
NACRES:
NB.03
€52.80
Check Cart for Availability
description
Standard Hub with 304 Stainless Steel Tubing
manufacturer/tradename
Cadence Science 4184
L
2 in.
gauge
20
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)