WHA1950002
Whatman® glass microfiber filtration funnel
Filter Funnel, 3-piece, volume 16 mL, diam. 25 mm
Synonym(s):
Whatman filter, Z741159, filter funnel, glass funnel
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
Filter Funnel, 3-piece
packaging
pkg of 1 ea
manufacturer/tradename
Whatman 1950-002
Whatman Article No. 28418621 (US reference)
diam.
25 mm
volume
16 mL
capacity
16 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Chemically resistant - 3-Piece construction consists of glass cylinder and base with a polymeric membrane support.
- Support options - Provided with acrylic support and optional polypropylene or PTFE supports available separately for some sizes.
- Secure sealing - Clamps at edges to prevent peripheral loss due to fluid bypass around the edge of the filter.
- Eliminate carry-over -speed of design facilitates easy and thorough cleaning between use.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service