Skip to Content
Merck
All Photos(1)

Key Documents

W523607

Sigma-Aldrich

Petitgrain oil, Paraguay

Synonym(s):

Bitter orange oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502

biological source

Citrus aurantium

grade

Kosher

optical activity

[α]20/D −3.0°, neat

refractive index

n20/D 1.459 (lit.)

bp

173 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

woody

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: steam distillation

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

152.0 °F

Flash Point(C)

66.67 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service