Skip to Content
Merck
All Photos(1)

Key Documents

W303704

Sigma-Aldrich

Styrax

FG

Synonym(s):

Gum storax, Styrax

Sign Into View Organizational & Contract Pricing

Select a Size

W303704-SAMPLE-K
€47.40
1 KG
€250.00
5 KG
€1,060.00
10 KG
€1,660.00

€47.40


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W303704-SAMPLE-K
€47.40
1 KG
€250.00
5 KG
€1,060.00
10 KG
€1,660.00

About This Item

CAS Number:
FEMA Number:
3037
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
Liquidambar spp.
food allergen:
no known allergens

€47.40


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

biological source

Liquidambar spp.

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

origin

Honduras origin

refractive index

n20/D 1.59 (lit.)

density

1.192 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsamic

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 15 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 104-104 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service