Skip to Content
Merck
All Photos(1)

Key Documents

W501506

Sigma-Aldrich

2-Furoic acid

≥97%

Synonym(s):

Furan-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4O3
CAS Number:
Molecular Weight:
112.08
Beilstein:
110149
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.136

Assay

≥97%

bp

230-232 °C (lit.)

mp

128-132 °C (lit.)

SMILES string

OC(=O)c1ccco1

InChI

1S/C5H4O3/c6-5(7)4-2-1-3-8-4/h1-3H,(H,6,7)

InChI key

SMNDYUVBFMFKNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Odorless

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

282.7 °F - closed cup

Flash Point(C)

139.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Yuan Li et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 31(5), 645-647 (2008-10-02)
The metabolites of a marine streptomyces sp. actinomycete (No. 195-02) were studied and eight compounds were isolated from the fermentation liquid, structures were elucidated by spectroscopy methods as p-hydroxy-benzonitrile (1), 2-methyl-furan-3-carboxylic acid(2), furan-2-carboxylic acid (3), cyclo(Phe-Phe) (4), cyclo(Leu-Ileu) (5), nicotinic
María C Rodríguez-Argüelles et al.
Journal of inorganic biochemistry, 103(1), 35-42 (2008-10-25)
Cobalt, nickel, copper and zinc coordination compounds of two thiosemicarbazones with general composition ML(2) (L: monodeprotonated ligand corresponding to 2-acetyl-gamma-butyrolactone thiosemicarbazone, HL(1), and 2-furancarbaldehyde thiosemicarbazone, HL(2)) and also complexes with general composition MCl(2)(HL(2)) were synthesized (except [NiCl(2)(HL(2))] and [Co(L(2))(2)]). The
Z B Tan et al.
Journal of analytical toxicology, 27(1), 43-46 (2003-02-18)
An improved high-performance liquid chromatographic (HPLC) method for the analysis of the metabolite furoic acid in the urine of workers occupationally exposed to furfural is described. The procedure involved an alkaline hydrolysis step followed by solvent extraction using ethyl acetate.
Hiroshi Yanagita et al.
Bioorganic & medicinal chemistry, 19(2), 816-825 (2011-01-05)
Rapid emergence of drug-resistant variants is one of the most serious problems in chemotherapy for HIV-1 infectious diseases. Inhibitors acting on a target not addressed by approved drugs are of great importance to suppress drug-resistant viruses. HIV-1 reverse transcriptase has
H B Klinke et al.
Applied microbiology and biotechnology, 57(5-6), 631-638 (2002-01-10)
Alkaline wet oxidation (WO) (using water, 6.5 g/l sodium carbonate, and 12 bar oxygen at 195 degrees C) was used for pre-treating wheat straw (60 g/l), resulting in a hemicellulose-rich hydrolysate and a cellulose-rich solid fraction. The hydrolysate consisted of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service