Merck
  • [Secondary metabolites of a marine actinomycete Streptomyces sp. (No. 195-02) from South China Sea].
[Secondary metabolites of a marine actinomycete Streptomyces sp. (No. 195-02) from South China Sea].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2008-10-02)
Chun-Yuan Li, Wei-Jia Ding, Zhi-Gang She, Yong-Cheng Lin
ABSTRACT
The metabolites of a marine streptomyces sp. actinomycete (No. 195-02) were studied and eight compounds were isolated from the fermentation liquid, structures were elucidated by spectroscopy methods as p-hydroxy-benzonitrile (1), 2-methyl-furan-3-carboxylic acid(2), furan-2-carboxylic acid (3), cyclo(Phe-Phe) (4), cyclo(Leu-Ileu) (5), nicotinic acid (6), 2-(1H-indol-3-yl) acetic acid (7) and bis(2-ethylhexyl) phthalate (8). The compounds 1, 3 and 8 were firstly isolated from Streptomyces sp., compounds 4 -7 were firstly found from marine actinomycetes. Coumpouds 4 and 5 evidently inhibited the growth of cancer cell lines hepG2 and hep2 at the concentration of 50 microg/ml.
Sigma-Aldrich
2-Furoic acid, 98%