Skip to Content
Merck
All Photos(1)

Key Documents

W394807

Sigma-Aldrich

4-Methylcyclohexanone

≥99%

Synonym(s):

Tetrahydro-p-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H9(=O)
CAS Number:
Molecular Weight:
112.17
FEMA Number:
3948
Beilstein:
506746
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

reg. compliance

FDA 21 CFR 117

Assay

≥99%

refractive index

n20/D 1.4443 (lit.)

bp

169-171 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

musty; animal

SMILES string

CC1CCC(=O)CC1

InChI

1S/C7H12O/c1-6-2-4-7(8)5-3-6/h6H,2-5H2,1H3

InChI key

VGVHNLRUAMRIEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methylcyclohexanone is a cyclic flavor ketone that is reported to be present in volatile oil of roasted sesame seeds.[1]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of volatile flavor components of the oil from roasted sesame seeds.
Nakamura S, et al.
Agricultural and Biological Chemistry, 53(7), 1891-1899 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service