Skip to Content
Merck
All Photos(1)

Key Documents

29140

Sigma-Aldrich

Cyclohexanone

puriss. p.a., ≥99.5% (GC)

Synonym(s):

pimelic ketone

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€40.80
500 ML
€53.30
1 L
€93.70

€40.80


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€40.80
500 ML
€53.30
1 L
€93.70

About This Item

Linear Formula:
C6H10(=O)
CAS Number:
Molecular Weight:
98.14
Beilstein:
385735
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
155 °C (lit.)
vapor pressure:
3.4 mmHg ( 20 °C)

€40.80


Estimated to ship on30 May 2025


Request a Bulk Order

vapor density

3.4 (vs air)

Quality Level

vapor pressure

3.4 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.5% (GC)

autoignition temp.

788 °F

expl. lim.

1.1 %, 100 °F
9.4 %

technique(s)

GC/GC: suitable

impurities

≤0.5% water

evapn. residue

≤0.05%

refractive index

n20/D 1.450 (lit.)
n20/D 1.450

bp

155 °C (lit.)

mp

−47 °C (lit.)

density

0.947 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

functional group

ketone

SMILES string

O=C1CCCCC1

InChI

1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2

InChI key

JHIVVAPYMSGYDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexanone is a cyclic ketone mainly used as an intermediate in the synthesis of nylon.[1]

Application

Cyclohexanone may undergo asymmetric Michael addition with aryl and alkyl nitroolefins in the presence of pyrrolidine-urea based catalysts to form the corresponding adducts.[2] It may also be used in the synthesis of a wide range of compounds such as adipic acid[3], 2-(hydroxy(phenyl)methyl)cyclohexanone[4], 2-allylcyclohexan-1-one[5], tetrasubstituted propargylamines[6] and N-cyclohexyl-benzyl amine derivatives.[7]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrrolidine- thiourea as a bifunctional organocatalyst: highly enantioselective Michael addition of cyclohexanone to nitroolefins
Cao CL, et al
Organic Letters, 8(14), 2901-2904 (2006)
(S)-(-)-2-allylcyclohexanone
Braun M, et al
Organic Syntheses, 86, 47-47 (2009)
First Order Hyperpolarizabilities, NPA and Fukui Functions of Cyclohexanone by Density Functional Theory Method.
Gangadharan RP and Krishnan SS.
Acta Physica Polonica A, 127(3), 748-752 (2015)
Conversion of Cyclohexanone to Adipic Acid Catalyzed by Heteropoly Compounds.
Lesbani A, et al.
Indonesian Journal of Chemistry, 15(1), 64-69 (2015)
194. A synthesis of vitamin a from cyclohexanone.
Attenburrow J, et al.
Journal of the Chemical Society, 1094-1111 (1952)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service