Skip to Content
Merck
All Photos(1)

Key Documents

W389404

Sigma-Aldrich

Phenylethyl mercaptan

≥99%, FG

Synonym(s):

2-Phenylethanethiol, 2-Phenylethyl mercaptan, Phenethyl mercaptan

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€71.20
100 G
€215.00
1 KG
€1,760.00

€71.20


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
25 G
€71.20
100 G
€215.00
1 KG
€1,760.00

About This Item

Linear Formula:
C6H5CH2CH2SH
CAS Number:
Molecular Weight:
138.23
FEMA Number:
3894
Beilstein:
1446618
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.194
NACRES:
NA.21
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

€71.20


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA

Assay

≥99%

refractive index

n20/D 1.560 (lit.)

bp

217-218 °C (lit.)

density

1.032 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sulfurous

SMILES string

SCCc1ccccc1

InChI

1S/C8H10S/c9-7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

ZMRFRBHYXOQLDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jishi Chen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(72), 18187-18192 (2017-10-17)
Double-crown Ni, Pd, or Pt nanoclusters have attracted extensive interests due to their aesthetic structure and intriguing properties. However, their doping by other metals remains unknown until now. Herein, Pd
Akhtar Munir et al.
ChemSusChem, 12(23), 5117-5125 (2019-10-28)
Metal nanoclusters (NCs, size ≤2 nm) are emerging materials in catalysis owing to their unique catalytic and electronic properties such as high surface/volume ratio, high redox potential, plethora of surface active sites, and dynamic behavior on a suitable support during catalysis.
Junying Zhang et al.
Nanoscale, 9(43), 16879-16886 (2017-10-28)
Size-dependence is an important factor in gold nanocatalysis. In this study, we explored the catalytic performance of atomically precise Au
Chao Liu et al.
ChemSusChem, 10(9), 1976-1980 (2017-03-30)
The catalytic performance of the atomically precise gold cluster-Au
Yingwei Li et al.
ACS nano, 14(6), 6599-6606 (2020-04-15)
Dipole moment (μ) is a critical parameter for molecules and nanomaterials as it affects many properties. In metal-thiolate (SR) nanoclusters (NCs), μ is commonly low (0-5 D) compared to quantum dots. Herein, we report a doping strategy to give giant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service