Merck
  • Efficient Aerobic Oxidation of Glucose to Gluconic Acid over Activated Carbon-Supported Gold Clusters.
Efficient Aerobic Oxidation of Glucose to Gluconic Acid over Activated Carbon-Supported Gold Clusters.
ChemSusChem (2017-03-30)
Chao Liu, Junying Zhang, Jiahui Huang, Chaolei Zhang, Feng Hong, Yang Zhou, Gao Li, Masatake Haruta
ABSTRACT
The catalytic performance of the atomically precise gold cluster-Au
Sigma-Aldrich
Phenylethyl mercaptan, ≥99%, FG
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis