Skip to Content
Merck
All Photos(1)

Key Documents

W387800

Sigma-Aldrich

Bis(methylthio)methane

≥99%, FG

Synonym(s):

Truffle sulfide, Dimethylthiomethane, Formaldehyde dimethyl mercaptal

Sign Into View Organizational & Contract Pricing

Select a Size

W387800-SAMPLE-K
€47.50
100 G
€103.00
1 KG
€700.00
2.5 KG
€779.00

€47.50


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W387800-SAMPLE-K
€47.50
100 G
€103.00
1 KG
€700.00
2.5 KG
€779.00

About This Item

Linear Formula:
CH3SCH2SCH3
CAS Number:
Molecular Weight:
108.23
FEMA Number:
3878
Beilstein:
1731143
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.118
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€47.50


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥99%

refractive index

n20/D 1.53 (lit.)

bp

147 °C (lit.)

density

1.059 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

garlic; green; mushroom; spicy; sulfurous

SMILES string

CSCSC

InChI

1S/C3H8S2/c1-4-3-5-2/h3H2,1-2H3

InChI key

LOCDPORVFVOGCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Natural occurrence: Truffles cheese, shittake mushrooms, and lobster.
Bis(methylthio)methane, a volatile sulfur compound, is the key component of white truffle aroma.[1] It is also reported to be an off-flavor in crustaceans such as prawn and sand lobster.[2]

Application


  • RIFM fragrance ingredient safety assessment, bis-(methylthio)methane, CAS Registry Number 1618-26-4.: This safety assessment explores the use of bis(methylthio)methane as a fragrance ingredient. The study evaluates its toxicological profile, confirming its safety for use in various cosmetic and personal care products (Api et al., 2021).

  • Characterization of the Key Aroma Compounds in Three Truffle Varieties from China by Flavoromics Approach.: Bis(methylthio)methane is identified as a significant aroma compound in Chinese truffle varieties. The research uses a flavoromics approach to map out the aroma profiles, supporting the enhancement of truffle-based products (Feng et al., 2019).

  • Multidimensional Gas Chromatography Coupled to Combustion-Isotope Ratio Mass Spectrometry/Quadrupole MS with a Low-Bleed Ionic Liquid Secondary Column for the Authentication of Truffles and Products Containing Truffle.: This analytical method leverages bis(methylthio)methane as a marker for authenticating truffle products. The study improves the reliability of truffle product verification, ensuring quality control in the food industry (Sciarrone et al., 2018).

  • Olfactometry Profiles and Quantitation of Volatile Sulfur Compounds of Swiss Tilsit Cheeses.: Bis(methylthio)methane is quantified as a key sulfur compound in Swiss Tilsit cheese. The study′s findings contribute to the understanding of cheese aroma profiles, aiding in the development of flavor-enhanced dairy products (Fuchsmann et al., 2015).


Biochem/physiol Actions

Odor at 0.1% PG
Taste at 0.10-0.30 ppm

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bis-(methylthio)-methane, an important off-flavour component in prawns and sand lobsters.
Whitfield FB, et al.
Chemistry & Industry, 158-159 (1981)
Headspace solid-phase microextraction analysis of volatile organic sulfur compounds in black and white truffle aroma.
Pelusio F, et al.
Journal of Agricultural and Food Chemistry, 43(8), 2138-2143 (1995)
Mosciano, G.
Perfumer & Flavorist, 26, 71-71 (2001)
Osvaldo Zarivi et al.
FEMS microbiology letters, 220(1), 81-88 (2003-03-20)
Tyrosinase is an enzyme having two copper atoms at the reactive site occurring in prokaryotic and eukaryotic organisms. In animals tyrosinase is responsible for pigmentation, in plants for protection of injured tissues or, as in fungi, to harden cell walls.
Mohamed Osman Awaleh et al.
Inorganic chemistry, 46(8), 3185-3191 (2007-03-17)
Two topologically comparable complexes, [Ag6(CF3CO2)3(L1-Me)3(SCH3)3]infinity (1) and [Ag6(CF3CF2CO2)3(L1-Me)2(SCH3)3(H2O)]infinity (2), were prepared and characterized by single-crystal diffractometry. The structures consist of Ag12S6 clusters linked by bis(methylthio)methane ligands, L1-Me, thus forming 1D coordination polymers. The 12 Ag atoms of the cluster are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service