Skip to Content
Merck
All Photos(1)

Key Documents

W348630

Sigma-Aldrich

Ethyl trans-2-butenoate

natural, ≥97%, FG

Synonym(s):

Ethyl crotonate

Sign Into View Organizational & Contract Pricing

Select a Size

W348630-SAMPLE-K
€52.30
500 G
€193.00
1 KG
€298.00

€52.30


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W348630-SAMPLE-K
€52.30
500 G
€193.00
1 KG
€298.00

About This Item

Linear Formula:
CH3CH=CHCOOC2H5
CAS Number:
FEMA Number:
3486
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.248
NACRES:
NA.21
grade:
FG
Kosher
natural
food allergen:
no known allergens

€52.30


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

FG
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.425

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; pungent; rum; sweet

SMILES string

O(CC)C(=O)\C=C\C

InChI

1S/C6H10O2/c1-3-5-6(7)8-4-2/h3,5H,4H2,1-2H3/b5-3+

InChI key

ZFDIRQKJPRINOQ-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengya Wang et al.
Applied microbiology and biotechnology, 102(19), 8511-8524 (2018-08-09)
Persisters represent a small bacterial population that is dormant and that survives under antibiotic treatment without experiencing genetic adaptation. Persisters are also considered one of the major reasons for recalcitrant chronic bacterial infections. Although several mechanisms of persister formation have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service