Skip to Content
Merck
All Photos(2)

Key Documents

W338820

Sigma-Aldrich

2-Tridecanone

≥98%, natural, FG

Synonym(s):

Methyl undecyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COCH3
CAS Number:
Molecular Weight:
198.34
FEMA Number:
3388
Beilstein:
1757402
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
7.103
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
botanical
food allergen:
no known allergens

biological source

botanical

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.510

Assay

≥98%

refractive index

n20/D 1.435 (lit.)

bp

134 °C/10 mmHg (lit.)
263 °C (lit.)

mp

24-27 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fatty; mushroom; waxy

SMILES string

CCCCCCCCCCCC(C)=O

InChI

1S/C13H26O/c1-3-4-5-6-7-8-9-10-11-12-13(2)14/h3-12H2,1-2H3

InChI key

CYIFVRUOHKNECG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service