Skip to Content
Merck
All Photos(1)

Key Documents

W320803

Sigma-Aldrich

(Methylthio)methylpyrazine

mixture of isomers, ≥98%, FG

Synonym(s):

Popcorn thiopyrazine

Sign Into View Organizational & Contract Pricing

Select a Size

W320803-SAMPLE-K
€47.40
100 G
€285.00
1 KG
€1,280.00

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W320803-SAMPLE-K
€47.40
100 G
€285.00
1 KG
€1,280.00

About This Item

Empirical Formula (Hill Notation):
C6H8N2S
CAS Number:
Molecular Weight:
140.21
FEMA Number:
3208
EC Number:
Council of Europe no.:
2290
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.035
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥98%

refractive index

n20/D 1.583 (lit.)

density

1.145 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cocoa; coffee; hazelnut; roasted

SMILES string

CSc1cnc(C)cn1.CSc2nccnc2C

InChI

1S/2C6H8N2S/c1-5-3-8-6(9-2)4-7-5;1-5-6(9-2)8-4-3-7-5/h2*3-4H,1-2H3

InChI key

PYWKOMLPQRZQSD-UHFFFAOYSA-N

General description

(Methylthio)methylpyrazine (mixture of isomers) are pyrazine compounds that are mainly formed in food products by Maillard reaction between sugars and proteins during cooking or roasting.[1] It occurs naturally in tobacco.[2]

Biochem/physiol Actions

Taste at 3-10 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

192.2 °F

Flash Point(C)

89 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Chemical Components of Tobacco and Tobacco Smoke (2016)
Flavours and Fragrances (1997)
Mosciano, G.
Perfumer & Flavorist, 25, 52-52 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service