Skip to Content
Merck
All Photos(1)

Key Documents

W257001

Sigma-Aldrich

Hexyl formate

≥97%, FG

Synonym(s):

Formic acid hexyl ester, Hexyl methanoate

Sign Into View Organizational & Contract Pricing

Select a Size

W257001-SAMPLE-K
€48.80
1 KG
€112.00
9 KG
€1,250.00
20 KG
€2,650.00

€48.80


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W257001-SAMPLE-K
€48.80
1 KG
€112.00
9 KG
€1,250.00
20 KG
€2,650.00

About This Item

Linear Formula:
HCO2(CH2)5CH3
CAS Number:
Molecular Weight:
130.18
FEMA Number:
2570
EC Number:
Council of Europe no.:
499
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.161
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€48.80


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.407 (lit.)

bp

155-156 °C (lit.)

mp

−63 °C (lit.)

density

0.879 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; ethereal; fruity

SMILES string

[H]C(=O)OCCCCCC

InChI

1S/C7H14O2/c1-2-3-4-5-6-9-7-8/h7H,2-6H2,1H3

InChI key

OUGPMNMLWKSBRI-UHFFFAOYSA-N

Related Categories

Application


  • Toxicities of 31 volatile low molecular weight compounds against Aedes aegypti and Culex quinquefasciatus.: Highlighting the practical applications of hexyl formate in public health, this study evaluates its efficacy as an insecticidal agent against mosquito species known for transmitting diseases. Such research is vital for developing environmentally friendly pest control methods (Chaskopoulou et al., 2009).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vivek Kempraj et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-06)
The green tree ant, Oecophylla smaragdina, is one of only two recognized species of weaver ants. While the identity and functions of chemicals produced and emitted by its congener O. longinoda have been studied quite extensively and serve as a
Rong Zhang et al.
Journal of agricultural and food chemistry, 62(33), 8302-8307 (2014-07-30)
Nonvolatile compounds play important roles in the quality of alcoholic beverages. In our previous work, a type of cyclooctapeptide lichenysin was newly identified in Chinese strong-aroma type liquor. In this work, it was found that lichenysin could selectively affect aroma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service