Skip to Content
Merck
All Photos(1)

Key Documents

W316814

Sigma-Aldrich

3,4-Hexanedione

≥95%, FG

Synonym(s):

Hexane-3,4-dione

Sign Into View Organizational & Contract Pricing

Select a Size

W316814-SAMPLE-K
€47.40
100 G
€117.00
250 G
€237.00
1 KG
€705.00
4 KG
€2,020.00

€47.40


Estimated to ship on24 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W316814-SAMPLE-K
€47.40
100 G
€117.00
250 G
€237.00
1 KG
€705.00
4 KG
€2,020.00

About This Item

Linear Formula:
CH2CH2COCOCH2CH3
CAS Number:
Molecular Weight:
114.14
FEMA Number:
3168
Beilstein:
1700837
EC Number:
Council of Europe no.:
2255
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.077
NACRES:
NA.21
Organoleptic:
caramel; buttery
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens

€47.40


Estimated to ship on24 March 2025


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥95%

refractive index

n20/D 1.41 (lit.)

bp

131 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caramel; buttery

SMILES string

CCC(=O)C(=O)CC

InChI

1S/C6H10O2/c1-3-5(7)6(8)4-2/h3-4H2,1-2H3

InChI key

KVFQMAZOBTXCAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

3,4-Hexanedione occurs naturally in honey[1] and coffee.[2] It has been proposed as a substitute for diacetyl in butter flavorings.[3]

Biochem/physiol Actions

Odor at 1.0%
Taste at 5 ppm

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Profiles of volatile compounds and sensory analysis of three blends of coffee: influence of different proportions of Arabica and Robusta and influence of roasting coffee with sugar.
Sanz C, et al.
Journal of the Science of Food and Agriculture, 82(8), 840-847 (2002)
Flavor and free amino acid composition of lavender and eucalyptus honeys.
Bouseta A, et al.
Journal of Food Science, 61(4), 683-687 (1996)
Stacey E Anderson et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 62, 373-381 (2013-09-07)
Concern has been raised over the association of diacetyl with lung disease clinically resembling bronchiolitis obliterans in food manufacturing workers. This has resulted in the need for identification of alternative chemicals to be used in the manufacturing process. Structurally similar
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30(5), 49-49 (2005)
Elizabeth I Parkinson et al.
Bioorganic & medicinal chemistry, 19(15), 4635-4643 (2011-07-08)
Carboxylesterases (CE) are ubiquitous enzymes found in both human and animal tissues and are responsible for the metabolism of xenobiotics. This includes numerous natural products, as well as a many clinically used drugs. Hence, the activity of these agents is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service