Skip to Content
Merck
All Photos(3)

Key Documents

B85307

Sigma-Aldrich

2,3-Butanedione

97%

Synonym(s):

Biacetyl, Diacetyl

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€47.50
100 ML
€92.00
500 ML
€147.00

€47.50


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€47.50
100 ML
€92.00
500 ML
€147.00

About This Item

Linear Formula:
CH3COCOCH3
CAS Number:
Molecular Weight:
86.09
Beilstein:
605398
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

€47.50


Estimated to ship on30 March 2025


Request a Bulk Order

vapor density

3 (vs air)

Quality Level

vapor pressure

52.2 mmHg ( 20 °C)

Assay

97%

form

liquid

autoignition temp.

345 °C

refractive index

n20/D 1.394 (lit.)

bp

88 °C (lit.)

density

0.981 g/mL (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)C(C)=O

InChI

1S/C4H6O2/c1-3(5)4(2)6/h1-2H3

InChI key

QSJXEFYPDANLFS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Inactivates aminopeptidase-N.[1] Cyclocondensation with amines has been used to form triazine[2] and pteridine ring systems.[3] Also used as a precursor to α-diones.[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Helene et al.
European journal of biochemistry, 196(2), 385-393 (1991-03-14)
Sequence analysis of aminopeptidase N has shown that this zinc exopeptidase contains a consensus sequence (Val-Xaa-Xaa-His-Glu-Xaa-Xaa-His), generally found at the active site of zinc endopeptidases [Jongeneel, C. V., Bouvier, J. and Bairoch, A. (1989) FEBS Lett. 242, 211-214]. This suggests
T R Leonard et al.
American journal of physiology. Cell physiology, 299(6), C1398-C1401 (2010-09-17)
It has been accepted for half a century that, for a given level of activation, the steady-state isometric force of a muscle sarcomere depends exclusively on the amount of overlap between the contractile filaments actin and myosin, or equivalently sarcomere
Jhonatan A Hernandez-Valdes et al.
Frontiers in microbiology, 11, 1032-1032 (2020-06-12)
Some secondary metabolites of fermentative bacteria are desired compounds for the food industry. Examples of these compounds are diacetyl and acetaldehyde, which are produced by species of the lactic acid bacteria (LAB) family. Diacetyl is an aromatic compound, giving the
Tetrahedron Letters, 32, 3879-3879 (1991)
Australian Journal of Chemistry, 43, 1449-1449 (1990)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service