Skip to Content
Merck
All Photos(1)

Key Documents

W270504

Sigma-Aldrich

Methyl heptanoate

≥99%, FG

Synonym(s):

Methyl enanthate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5COOCH3
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2705
Beilstein:
1747147
EC Number:
Council of Europe no.:
368
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.096
NACRES:
NA.21
Organoleptic:
green; fruity; sweet
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

≥99%

refractive index

n20/D 1.411 (lit.)

bp

172-173 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity; sweet

SMILES string

CCCCCCC(=O)OC

InChI

1S/C8H16O2/c1-3-4-5-6-7-8(9)10-2/h3-7H2,1-2H3

InChI key

XNCNNDVCAUWAIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl heptanoate has been identified in pineapple, Hardy kiwifruit and salak fruit.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of edible coating on volatile compounds of hardy kiwifruit during storage.
ACS Symp. Ser., 1035, 79?94-79?94 (2010)
Volatile constituents of salak (Salacca edulis Reinw.) fruit.
Wong KC & Tie DY.
Flavour and Fragrance Journal, 8(6), 321-324 (1993)
Volatile constituents of pineapple (Ananas comosus [L.] Merr.)
Takeoka G, et al.
ACS Symp. Ser., 388, 223?237-223?237 (1989)
Silawan Somboonchan et al.
Food chemistry, 195, 79-86 (2015-11-18)
The effect of modulating the gelatinisation extent by hydration (50/50 and 80/20 water to starch ratio) and temperature (65 or 85 °C) on various properties of wheat starch in presence of flavours has been studied. The hydrothermal treatments resulted in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service