Skip to Content
Merck
All Photos(1)

Key Documents

W221805

Sigma-Aldrich

Butyl isovalerate

≥98%, FCC, FG

Synonym(s):

Butyl 3-methyl butyrate, Butyl 3-methylbutanoate, Iso valeric acid butyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

W221805-SAMPLE-K
€47.40
1 KG
€78.50
4 KG
€193.00
20 KG
€628.00

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W221805-SAMPLE-K
€47.40
1 KG
€78.50
4 KG
€193.00
20 KG
€628.00

About This Item

Linear Formula:
(CH3)2CHCH2COO(CH2)3CH3
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2218
Beilstein:
1752803
EC Number:
Council of Europe no.:
444
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.449
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.409 (lit.)

bp

175 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; green; fruity; peach; pineapple; sweet

SMILES string

CCCCOC(=O)CC(C)C

InChI

1S/C9H18O2/c1-4-5-6-11-9(10)7-8(2)3/h8H,4-7H2,1-3H3

InChI key

AYWJSCLAAPJZEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Butyl isovalerate is one of the volatile compounds found in jackfruit. It is responsible for the sweet and fruity flavor of jackfruit.[1][2]

Application


  • Effect of volatile organic compounds from bacteria on nematodes.: This research explores the impact of bacterial volatile organic compounds, including butyl isovalerate, on nematodes. The study reveals significant biological interactions and suggests applications in biocontrol and agricultural practices to manage nematode populations (Xu et al., 2015).

Biochem/physiol Actions

Taste at 10 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aroma volatiles from two fruit varieties of jackfruit (Artocarpus heterophyllus Lam.)
Maia JGS, et al.
Food Chemistry, 85, 195-197 (2004)
Analysis of volatile compounds in five jackfruit (Artocarpus heterophyllus L.) cultivars using solid-phase microextraction (SPME) and gas chromatography-time-of-flight mass spectrometry (GC-TOFMS)
Ong BT, et al.
J. Food Compos. Anal., 21, 416-422 (2008)
Mosciano, G.
Perfumer & Flavorist, 24, 49-49 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service