Skip to Content
Merck
All Photos(3)

Key Documents

T82201

Sigma-Aldrich

Triphenylborane

powder, <2% H2O

Synonym(s):

Triphenylborine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3B
CAS Number:
Molecular Weight:
242.12
Beilstein:
1961313
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder
solid

impurities

<2% H2O

bp

203 °C/15 mmHg (lit.)

mp

145 °C (lit.)

SMILES string

c1ccc(cc1)B(c2ccccc2)c3ccccc3

InChI

1S/C18H15B/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

MXSVLWZRHLXFKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triphenylborane powder is a boron based precursor that can be used in the chemical vapor deposition for the development of carbon nanomaterials for application in oxygen reduction reaction, hydrogen storage, transparent conducting films, and fluoride shuffle batteries.[1][2][3]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Boron-doped carbon nanotubes as metal-free electrocatalysts for the oxygen reduction reaction
Yang L, et al.
Angewandte Chemie (International ed. in English), 50(31), 7132-7135 (2011)
Metal-free doped carbon materials as electrocatalysts for the oxygen reduction reaction
Daems N, et al.
Journal of Material Chemistry A, 2(12), 4085-4110 (2014)
Triphenylboroxine and triphenylborane as anion acceptors for electrolyte in fluoride shuttle batteries
Konishi H, et al.
Chemistry Letters (Jpn), 47(11), 1346-1349 (2018)

Articles

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service