Skip to Content
Merck
All Photos(1)

Key Documents

418714

Sigma-Aldrich

Triphenyl borate

≥97%

Synonym(s):

NSC 806, Triphenoxyborane, Triphenyl ester boric acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€72.80

€72.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€72.80

About This Item

Linear Formula:
(C6H5O)3B
CAS Number:
Molecular Weight:
290.12
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€72.80


Check Cart for Availability

Request a Bulk Order

Assay

≥97%

form

solid

mp

98-101 °C (lit.)

SMILES string

O(B(Oc1ccccc1)Oc2ccccc2)c3ccccc3

InChI

1S/C18H15BO3/c1-4-10-16(11-5-1)20-19(21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

MDCWDBMBZLORER-UHFFFAOYSA-N

Application

Catalyst involved in:
  • Multicomponent aziridination of aldehydes
  • Hetero-Diels-Alder reactions and Mukaiyama aldol condensation
  • Cross-metathesis of alkenes with 3-nitropropene
  • Asymmetric Claisen rearrangement of unactivated allyl vinyl ethers
  • Ethoxylation
  • Asymmetric aziridination of imines

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service