Skip to Content
Merck
All Photos(1)

Documents

T76201

Sigma-Aldrich

Trimethylene oxide

97%

Synonym(s):

1,3-Epoxypropane, 1,3-Propylene oxide, Cyclooxabutane, Oxacyclobutane, Oxetane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
Beilstein:
102382
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

5.09 psi ( 20 °C)

Quality Level

Assay

97%

refractive index

n20/D 1.388-1.396
n20/D 1.392 (lit.)

bp

50 °C (lit.)

density

0.893 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1COC1

InChI

1S/C3H6O/c1-2-4-3-1/h1-3H2

InChI key

AHHWIHXENZJRFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Trimethylene oxide is a strained cyclic ether which can be used to replace carbonyl / gem-dimethyl groups during molecular modelling of bioactive molecules.
  • It can be incorporated in various bioactive molecule and drug substances for its improved physicochemical properties, via novel C-H activation methods.
  • Trimethylene oxide can also be used in the synthesis of poly(trimethylene oxide) and related polymer electrolytes.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of polar side chains in Li+ coordination and transport properties of polyoxetane-based polymer electrolytes.
Sai R, et al.
Physical Chemistry Chemical Physics, 19(7), 5185-5194 (2017)
Synthesis and characterization of high molecular weight poly (trimethylene oxide).
Qureshi MY and Ochel M
Eur. Polymer J., 32(6), 691-693 (1996)
Oxetanes: recent advances in synthesis, reactivity, and medicinal chemistry.
Bull JA, et al.
Chemical Reviews, 116(19), 12150-12233 (2016)
Cross-linked poly (oxetane) matrix for polymer electrolyte containing lithium ions.
Tsutsumi H, et al.
Solid State Ionics, 262, 761-764 (2014)
Dong-Dong Wu et al.
Organic & biomolecular chemistry, 10(18), 3626-3635 (2012-04-13)
Photoinduced reactions of isatin and N-methyl-1,3,4-isoquinolinetrione with bicycloalkylidenes such as bicyclopropylidene, cyclopropylidenecyclobutane, cyclopropylidenecyclohexane and bicyclohexylidene were investigated. The reactions gave spirooxetanes as the major products derived from the [2 + 2] photocycloaddition pathway via 1,4-biradical recombination. Unusual products including the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service