Skip to Content
Merck
All Photos(1)

Key Documents

733296

Sigma-Aldrich

3-Hydroxyoxetane

95%

Synonym(s):

1,3-Epoxy-2-propanol, 3-Oxetanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€26.80

€26.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€26.80

About This Item

Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€26.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

availability

not available in Japan, EU

refractive index

n20/D 1.438

density

1.167 g/mL at 25 °C

functional group

ether
hydroxyl

SMILES string

OC1COC1

InChI

1S/C3H6O2/c4-3-1-5-2-3/h3-4H,1-2H2

InChI key

QMLWSAXEQSBAAQ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

189.0 °F

Flash Point(C)

87.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service