Skip to Content
Merck
All Photos(1)

Key Documents

T57703

Sigma-Aldrich

1-Tridecene

96%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€70.20

€70.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€70.20

About This Item

Linear Formula:
CH3(CH2)10CH=CH2
CAS Number:
Molecular Weight:
182.35
Beilstein:
1744660
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€70.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

refractive index

n20/D 1.432 (lit.)

bp

232-233 °C (lit.)

mp

−23 °C (lit.)

density

0.766 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC=C

InChI

1S/C13H26/c1-3-5-7-9-11-13-12-10-8-6-4-2/h3H,1,4-13H2,2H3

InChI key

VQOXUMQBYILCKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

[Substance toxicity and danger].
L V Mel'nikova
Gigiena i sanitariia, (1)(1), 94-94 (1981-01-01)
Qing-He Zhang et al.
Journal of chemical ecology, 30(9), 1849-1870 (2004-12-14)
Gas chromatographic-electroantennographic detection (G3C-EAD) experiments showed that antennae of males and females of the goldeneyed lacewing, Chrysopa oculata Say (Co. = Chrysopa), consistently responded to four compounds extracted from the abdominal cuticle of males:nonanal, nonanol, nonanoic acid, and (1R*,2S*,5R*,8R*)-iridodial. These
Sven Geiselhardt et al.
Die Naturwissenschaften, 95(3), 247-251 (2007-09-28)
Males of the genus Parastizopus (Coleoptera: Tenebrionidae) exhibit a special pheromone-emitting behaviour. They do a headstand, expose the aedeagus and remain in this posture for a few seconds. The pheromone emitted by P. transgariepinus was collected by solid-phase micro-extraction (100

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service