Skip to Content
Merck
All Photos(1)

Key Documents

N30404

Sigma-Aldrich

1-Nonene

96%

Synonym(s):

α-Nonene, 1-Nonylene, 1-n-Nonene, n-Non-1-ene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€141.00
25 G
€380.00

€141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€141.00
25 G
€380.00

About This Item

Linear Formula:
CH3(CH2)6CH=CH2
CAS Number:
Molecular Weight:
126.24
Beilstein:
1698439
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€141.00


Check Cart for Availability

Request a Bulk Order

vapor density

4.35 (vs air)

Quality Level

vapor pressure

11 mmHg ( 37.7 °C)

Assay

96%

form

liquid

refractive index

n20/D 1.416 (lit.)

bp

146 °C (lit.)

mp

−81 °C (lit.)

density

0.73 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC=C

InChI

1S/C9H18/c1-3-5-7-9-8-6-4-2/h3H,1,4-9H2,2H3

InChI key

JRZJOMJEPLMPRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Ian Price et al.
Journal of theoretical biology, 374, 83-93 (2015-04-07)
Mortality from influenza infections continues as a global public health issue, with the host inflammatory response contributing to fatalities related to the primary infection. Based on Ordinary Differential Equation (ODE) formalism, a computational model was developed for the in-host response

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service